siRNA Details
| Gene Symbol: Gm32172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK075633.1 | - | 3 | 609 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV010166.2 | - | 3 | 625 | Chr. 10 - 13871281 - 13879204 [+] on Build GRCm38 | |

Gm32172
Genomic Map