siRNA Details
| Gene Symbol: Pcgf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079197.1 | - | 1 | 901 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB110239.2 | - | 1 | 85 | Chr. 19 - 36459807 - 36460502 [+] on Build GRCm38 | |

Pcgf5
Genomic Map