siRNA Details
| Gene Symbol: 1700081N11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006973.1 | - | 2 | 398 | ||
| AV205735.1 | - | 1 | 204 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707027.1 | - | 2 | 398 | Chr. 12 - 54862991 - 54863664 [+] on Build GRCm38 | |

1700081N11Rik
Genomic Map