siRNA Details
| Gene Symbol: 1700021J08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006209.1 | - | 1 | 123 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706301.1 | - | 1 | 123 | Chr. 4 - 117002166 - 117003012 [-] on Build GRCm38 | |

1700021J08Rik
Genomic Map