siRNA Details
| Gene Symbol: Hhipl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005401.1 | - | 48 | |||
| AK129450.1 | - | 3227 | ||||
| AK166269.1 | - | 3708 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hhipl1| Gene Symbol: Hhipl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005401.1 | - | 48 | |||
| AK129450.1 | - | 3227 | ||||
| AK166269.1 | - | 3708 | ||||