siRNA Details
| Gene Symbol: 2010110E17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008378.1 | - | 1 | 22 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY708292.1 | - | 1 | 22 | Chr. 8 - 86885688 - 86886541 [+] on Build GRCm38 | |

2010110E17Rik
Genomic Map