siRNA Details
| Gene Symbol: 4930404O17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015089.1 | - | 1 | 67 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714618.1 | - | 1 | 64 | Chr. 14 - 124730376 - 124742795 [-] on Build GRCm38 | |

4930404O17Rik
Genomic Map