siRNA Details
Gene Symbol: 4930404O17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015089.1 | - | 2 | 533 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714618.1 | - | 2 | 530 | Chr. 14 - 124730376 - 124742795 [-] on Build GRCm38 |
