siRNA Details
| Gene Symbol: Gm17750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082072.1 | - | 1 | 40 | ||
| BE862222.1 | - | 1 | 42 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB595923.1 | - | 1 | 40 | Chr. 13 - 84025556 - 84064772 [-] on Build GRCm38 | |

Gm17750
Genomic Map