siRNA Details
| Gene Symbol: Gm40289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030193.1 | - | 2 | 426 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB617074.1 | - | 2 | 423 | Chr. 5 - 37983693 - 37984454 [+] on Build GRCm38 | |

Gm40289
Genomic Map