siRNA Details
| Gene Symbol: 2210414L08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019066.1 | - | 1 | 32 | ||
| AK131897.1 | - | 1 | 32 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2210414L08Rik| Gene Symbol: 2210414L08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019066.1 | - | 1 | 32 | ||
| AK131897.1 | - | 1 | 32 | |||