siRNA Details
| Gene Symbol: Tmem181b-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033520.1 | 1 | 1161 | |||
| GenBank mRNA | AK137412.1 | - | 1 | 1161 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK089688.1 | - | 475 | Chr. - - [-] on Build | ||

Tmem181b-ps
Genomic Map