siRNA Details
Gene Symbol: Gm31508 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133621.1 | - | 1275 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm31508 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133621.1 | - | 1275 |