siRNA Details
| Gene Symbol: 2600001M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA260145.1 | - | 1 | 237 | ||
| AK011139.1 | - | 1 | 939 | |||
| BG076636.2 | - | 1 | 19 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY467931.1 | - | 1 | 29 | Chr. 2 - 5870257 - 5870814 [+] on Build GRCm38 | |

2600001M11Rik
Genomic Map