siRNA Details
| Gene Symbol: Erbb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK160975.1 | - | 1663 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erbb2| Gene Symbol: Erbb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK160975.1 | - | 1663 | |||