siRNA Details
| Gene Symbol: Gm13093 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK081620.1 | - | 538 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13093| Gene Symbol: Gm13093 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK081620.1 | - | 538 | |||