siRNA Details
| Gene Symbol: Gm31786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029624.1 | - | 2, 3 | 157 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB615021.1 | - | 2, 3 | 157 | Chr. 8 - 47744707 - 47749662 [-] on Build GRCm38 | |

Gm31786
Genomic Map