siRNA Details
| Gene Symbol: A730070K21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043208.1 | - | 1 | 1135 | ||
| AL022774.1 | - | 1 | 68 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB255903.2 | - | 1 | 434 | Chr. 8 - 73199264 - 73199954 [-] on Build GRCm38 | |

A730070K21Rik
Genomic Map