siRNA Details
| Gene Symbol: A730070K21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043208.1 | - | 1 | 1238 | ||
| AL022774.1 | - | 1 | 171 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A730070K21Rik| Gene Symbol: A730070K21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043208.1 | - | 1 | 1238 | ||
| AL022774.1 | - | 1 | 171 | |||