siRNA Details
| Gene Symbol: Pantr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC094545.1 | - | 1435 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pantr1| Gene Symbol: Pantr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC094545.1 | - | 1435 | |||