siRNA Details
| Gene Symbol: Gm16845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040406.1 | 4 | 1040 | |||
| NR_040407.1 | 3 | 976 | ||||
| GenBank mRNA | AK154427.1 | - | 3 | 946 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16845| Gene Symbol: Gm16845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040406.1 | 4 | 1040 | |||
| NR_040407.1 | 3 | 976 | ||||
| GenBank mRNA | AK154427.1 | - | 3 | 946 | ||