siRNA Details
| Gene Symbol: Gm16845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040406.1 | 3 | 261 | |||
| NR_040407.1 | 2 | 197 | ||||
| GenBank mRNA | AA021721.1 | - | 3 | 261 | ||
| AI509114.1 | - | 3 | 261 | |||
| AK154427.1 | - | 2 | 167 | |||
| BY232108.1 | - | 2 | 183 | |||

Gm16845
Genomic Map