siRNA Details
| Gene Symbol: Gm14285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034299.1 | - | 1 | 1426 | ||
| BB082380.2 | - | 1 | 278 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE647611.1 | - | 1 | 27 | Chr. 2 - 131562766 - 131563148 [+] on Build GRCm38 | |

Gm14285
Genomic Map