siRNA Details
| Gene Symbol: Gm14285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034299.1 | - | 1 | 1547 | ||
| BB082380.2 | - | 1 | 399 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14285| Gene Symbol: Gm14285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034299.1 | - | 1 | 1547 | ||
| BB082380.2 | - | 1 | 399 | |||