siRNA Details
| Gene Symbol: 2700057C20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012449.1 | - | 1 | 13 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY711583.1 | - | 11 | Chr. 7 - 12498752 - 12499646 [+] on Build GRCm38 | ||

2700057C20Rik
Genomic Map