siRNA Details
Gene Symbol: Ints6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013790.1 | - | 1 | 517 | ||
AK049304.1 | - | 1 | 2127 | |||
AK169725.1 | - | 1 | 2078 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ints6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013790.1 | - | 1 | 517 | ||
AK049304.1 | - | 1 | 2127 | |||
AK169725.1 | - | 1 | 2078 |