siRNA Details
| Gene Symbol: Ints6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013790.1 | - | 1 | 918 | ||
| AK049304.1 | - | 1 | 2529 | |||
| AK169725.1 | - | 1 | 2480 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ints6| Gene Symbol: Ints6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013790.1 | - | 1 | 918 | ||
| AK049304.1 | - | 1 | 2529 | |||
| AK169725.1 | - | 1 | 2480 | |||