siRNA Details
| Gene Symbol: 4930518P08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045364.1 | 2 | 351 | |||
| GenBank mRNA | AK015835.1 | - | 2 | 353 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930518P08Rik| Gene Symbol: 4930518P08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045364.1 | 2 | 351 | |||
| GenBank mRNA | AK015835.1 | - | 2 | 353 | ||