siRNA Details
| Gene Symbol: Gm36173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136704.1 | - | 2 | 356 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721096.1 | - | 2 | 346 | Chr. 10 - 53235187 - 53260247 [+] on Build GRCm38 | |

Gm36173
Genomic Map