siRNA Details
| Gene Symbol: Gm13857 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133155.1 | - | 351 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ047467.1 | - | 2, 3 | 351 | Chr. 6 - 34421041 - 34445435 [-] on Build GRCm38 | |

Gm13857
Genomic Map