siRNA Details
| Gene Symbol: Kcnq1ot1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042559.1 | - | 1 | 336 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB590925.2 | - | 1 | 334 | Chr. 7 - 143248996 - 143249618 [-] on Build GRCm38 | |

Kcnq1ot1
Genomic Map