siRNA Details
| Gene Symbol: 4930548J01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045462.1 | 3 | 617 | |||
| GenBank mRNA | AK019751.1 | - | 3 | 618 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930548J01Rik| Gene Symbol: 4930548J01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045462.1 | 3 | 617 | |||
| GenBank mRNA | AK019751.1 | - | 3 | 618 | ||