siRNA Details
| Gene Symbol: Csnk1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042000.1 | - | 1 | 3133 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB234840.2 | - | 1 | 355 | Chr. 11 - 120975965 - 120976536 [-] on Build GRCm38 | |

Csnk1d
Genomic Map