siRNA Details
Gene Symbol: 1700030M09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045903.1 | 1 | 55 | |||
GenBank mRNA | AK006561.1 | - | 1 | 56 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706573.1 | - | 1 | 56 | Chr. 8 - 121544388 - 121547388 [+] on Build GRCm38 |
