siRNA Details
| Gene Symbol: 1700030M09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045903.1 | 3 | 671 | |||
| GenBank mRNA | AK006561.1 | - | 3 | 672 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706573.1 | - | 3 | 672 | Chr. 8 - 121544388 - 121547388 [+] on Build GRCm38 | |

1700030M09Rik
Genomic Map