siRNA Details
| Gene Symbol: Ppp2r5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034872.1 | - | 1 | 1623 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp2r5c| Gene Symbol: Ppp2r5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034872.1 | - | 1 | 1623 | ||