siRNA Details
| Gene Symbol: Gm3161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK090034.1 | - | 238 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm3161| Gene Symbol: Gm3161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK090034.1 | - | 238 | |||