siRNA Details
Gene Symbol: Gm38410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035476.1 | - | 3 | 411 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB627990.1 | - | 3 | 410 | Chr. 18 - 24339366 - 24342136 [+] on Build GRCm38 |
