siRNA Details
| Gene Symbol: 1700044K19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006697.1 | - | 1 | 356 | ||
| AV045425.2 | - | 1 | 154 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700044K19Rik| Gene Symbol: 1700044K19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006697.1 | - | 1 | 356 | ||
| AV045425.2 | - | 1 | 154 | |||