siRNA Details
| Gene Symbol: 1700044C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045624.1 | 6 | 816 | |||
| GenBank mRNA | AK006694.1 | - | 6 | 817 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700044C05Rik| Gene Symbol: 1700044C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045624.1 | 6 | 816 | |||
| GenBank mRNA | AK006694.1 | - | 6 | 817 | ||