siRNA Details
| Gene Symbol: 1700044C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045624.1 | 4 | 396 | |||
| GenBank mRNA | AK006694.1 | - | 4 | 397 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706686.1 | - | 4 | 398 | Chr. 14 - 118365878 - 118398170 [+] on Build GRCm38 | |

1700044C05Rik
Genomic Map