siRNA Details
| Gene Symbol: 1700042O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006691.1 | - | 1 | 829 | ||
| AK030060.1 | - | 1 | 825 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706682.1 | - | 1 | 826 | Chr. 3 - 8743680 - 8744557 [+] on Build GRCm38 | |

1700042O13Rik
Genomic Map