siRNA Details
| Gene Symbol: Gm5086 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046157.1 | 3 | 966 | |||
| GenBank mRNA | AK045784.1 | - | 3 | 967 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5086| Gene Symbol: Gm5086 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046157.1 | 3 | 966 | |||
| GenBank mRNA | AK045784.1 | - | 3 | 967 | ||