siRNA Details
| Gene Symbol: D530014G21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021287.1 | - | 1 | 58 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY730571.1 | - | 1 | 55 | Chr. 8 - 8488304 - 8495535 [+] on Build GRCm38 | |

D530014G21Rik
Genomic Map