siRNA Details
| Gene Symbol: Gpatch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017966.1 | - | 1 | 784 | ||
| AK142303.1 | - | 1 | 75 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpatch2| Gene Symbol: Gpatch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017966.1 | - | 1 | 784 | ||
| AK142303.1 | - | 1 | 75 | |||