siRNA Details
| Gene Symbol: 2310079F09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK010214.1 | - | 1 | 952 | ||
| AK142062.1 | - | 2 | 2847 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310079F09Rik| Gene Symbol: 2310079F09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK010214.1 | - | 1 | 952 | ||
| AK142062.1 | - | 2 | 2847 | |||