siRNA Details
| Gene Symbol: C030041B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021138.1 | - | 1 | 433 | ||
| AK044777.1 | - | 1 | 434 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY728282.1 | - | 1 | 431 | Chr. 6 - 32954284 - 32954945 [-] on Build GRCm38 | |

C030041B07Rik
Genomic Map