siRNA Details
| Gene Symbol: Gm17747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK146230.1 | - | 2 | 420 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY758231.1 | - | 2 | 420 | Chr. 14 - 25441641 - 25453223 [+] on Build GRCm38 | |

Gm17747
Genomic Map