siRNA Details
| Gene Symbol: Gm16308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK140961.1 | - | 438 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB376283.2 | - | 2 | 101 | Chr. 15 - 71810452 - 71813780 [+] on Build GRCm38 | |

Gm16308
Genomic Map