siRNA Details
Gene Symbol: Gm16308 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK140961.1 | - | 636 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB376283.2 | - | 3, 4 | 299 | Chr. 15 - 71810452 - 71813780 [+] on Build GRCm38 |
